Anti-FOXC1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FOXC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-341
- 77823-340
- Antigen symbol:
- FOXC1
- FOXC1
- Antigen name:
- Forkhead box protein C1 protein (1-79AA)
- Forkhead box protein C1 protein (1-79AA)
- Antigen synonyms:
- ARA|FKH L7|FKHL 7|FKHL7|Forkhead (Drosophila) like 7|Forkhead box C1|Forkhead box protein C1|Forkhead drosophila homolog like 7|Forkhead like 7|Forkhead related activator 3|Forkhead related protein FKHL7|Forkhead related transcription factor 3|Forkhead-related protein FKHL7|Forkhead-related transcription factor 3|FOX C1|FOXC 1|Foxc1|FOXC1_HUMAN|FREAC 3|FREAC-3|FREAC3|IGDA|IHG 1|IHG1|IRID 1|IRID1|Iridogoniodysgenesis type 1|Myeloid factor delta
- ARA|FKH L7|FKHL 7|FKHL7|Forkhead (Drosophila) like 7|Forkhead box C1|Forkhead box protein C1|Forkhead drosophila homolog like 7|Forkhead like 7|Forkhead related activator 3|Forkhead related protein FKHL7|Forkhead related transcription factor 3|Forkhead-related protein FKHL7|Forkhead-related transcription factor 3|FOX C1|FOXC 1|Foxc1|FOXC1_HUMAN|FREAC 3|FREAC-3|FREAC3|IGDA|IHG 1|IHG1|IRID 1|IRID1|Iridogoniodysgenesis type 1|Myeloid factor delta
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12948
- Q12948
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together