Anti-nef Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: nef
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human immunodeficiency virus 1
Specifications
- Catalog No:
- 77813-350
- 77813-349
- Antigen symbol:
- nef
- nef
- Antigen name:
- immunodeficiency virus 1 Protein Nef protein (1-209AA)
- immunodeficiency virus 1 Protein Nef protein (1-209AA)
- Antigen synonyms:
- Protein Nef, 3'ORF, Negative factor, F-protein, C-terminal core protein, nef
- Protein Nef, 3'ORF, Negative factor, F-protein, C-terminal core protein, nef
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human immunodeficiency virus 1
- Human immunodeficiency virus 1
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A0A1L4CSJ1
- A0A1L4CSJ1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together