Anti-SART3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SART3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-401
- 77825-400
- Antigen symbol:
- DSAP1;hSART 3;hSART-3;P100;P110;p110(nrb);RP11 13G14;SART 3;SART-3;SART3;SART3_HUMAN;Squamous cell carcinoma antigen recognized by T cells 3;Squamous cell carcinoma antigen recognized by T-cells 3;Tat interacting protein of 110 kDa;Tat-interacting protein of 110 kDa;Tip110
- DSAP1;hSART 3;hSART-3;P100;P110;p110(nrb);RP11 13G14;SART 3;SART-3;SART3;SART3_HUMAN;Squamous cell carcinoma antigen recognized by T cells 3;Squamous cell carcinoma antigen recognized by T-cells 3;Tat interacting protein of 110 kDa;Tat-interacting protein of 110 kDa;Tip110
- Antigen name:
- Squamous cell carcinoma antigen recognized by T-cells 3 protein (600-900AA)
- Squamous cell carcinoma antigen recognized by T-cells 3 protein (600-900AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15020
- Q15020
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together