Anti-XIAP Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: XIAP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-714
- 77804-713
- Antigen symbol:
- XIAP
- XIAP
- Antigen name:
- E3 ubiquitin-protein ligase XIAP protein (1-497AA)
- E3 ubiquitin-protein ligase XIAP protein (1-497AA)
- Antigen synonyms:
- AP 13|API3|Apoptosis Inhibitor 3|Baculoviral IAP repeat containing 4|Baculoviral IAP Repeat Containing Protein 4|Baculoviral IAP repeat-containing protein 4|BIRC 4|BIRC4|E3 ubiquitin-protein ligase XIAP|hIAP-3|hIAP3|HILP|IAP 3|IAP like protein|IAP-3|IAP-like protein|IAP3|ILP 1|ILP|ILP1|Inhibitor of apoptosis protein 3|Inhibitor of Apoptosis X Linked|Mammalian IAP Homologue A|MIHA|X linked IAP|X linked inhibitor of apoptosis|X linked inhibitor of apoptosis E3 ubiquitin protein ligase|X linked inhibitor of apoptosis protein|X-linked IAP|x-linked inhibitor of apopotosis proteins|X-linked inhibitor of apoptosis protein|Xiap|XIAP_HUMAN|XLP2
- AP 13|API3|Apoptosis Inhibitor 3|Baculoviral IAP repeat containing 4|Baculoviral IAP Repeat Containing Protein 4|Baculoviral IAP repeat-containing protein 4|BIRC 4|BIRC4|E3 ubiquitin-protein ligase XIAP|hIAP-3|hIAP3|HILP|IAP 3|IAP like protein|IAP-3|IAP-like protein|IAP3|ILP 1|ILP|ILP1|Inhibitor of apoptosis protein 3|Inhibitor of Apoptosis X Linked|Mammalian IAP Homologue A|MIHA|X linked IAP|X linked inhibitor of apoptosis|X linked inhibitor of apoptosis E3 ubiquitin protein ligase|X linked inhibitor of apoptosis protein|X-linked IAP|x-linked inhibitor of apopotosis proteins|X-linked inhibitor of apoptosis protein|Xiap|XIAP_HUMAN|XLP2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P98170
- P98170
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together