Anti-SLC24A1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SLC24A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-868
- 77799-869
- Antigen symbol:
- SLC24A1
- SLC24A1
- Antigen name:
- Sodium/potassium/calcium exchanger 1 protein (330-452AA)
- Sodium/potassium/calcium exchanger 1 protein (330-452AA)
- Antigen synonyms:
- CSNB1D|HsT17412|KIAA0702|Na(+)/K(+)/Ca(2+) exchange protein 1|Na(+)/K(+)/Ca(2+)-exchange protein 1|NCKX|NCKX1|NCKX1_HUMAN|Retinal rod Na Ca+K exchanger|Retinal rod Na+/Ca+/K+ exchanger|Retinal rod Na-Ca+K exchanger|RODX|Sodium/potassium/calcium exchanger 1|Solute carrier family 24 (sodium/potassium/calcium exchanger) member 1|Solute carrier family 24 member 1
- CSNB1D|HsT17412|KIAA0702|Na(+)/K(+)/Ca(2+) exchange protein 1|Na(+)/K(+)/Ca(2+)-exchange protein 1|NCKX|NCKX1|NCKX1_HUMAN|Retinal rod Na Ca+K exchanger|Retinal rod Na+/Ca+/K+ exchanger|Retinal rod Na-Ca+K exchanger|RODX|Sodium/potassium/calcium exchanger 1|Solute carrier family 24 (sodium/potassium/calcium exchanger) member 1|Solute carrier family 24 member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60721
- O60721
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together