Anti-STK32A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: STK32A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77813-342
- 77813-343
- Antigen symbol:
- STK32A
- STK32A
- Antigen name:
- Synthetic peptide of STK32A
- Synthetic peptide of STK32A
- Antigen synonyms:
- A930015B13Rik|MGC22688|Serine/threonine kinase 32A|Serine/threonine protein kinase 32A|Serine/threonine-protein kinase 32A|ST32A_HUMAN|STK 32A|YANK 1|YANK1|Yet another novel kinase 1
- A930015B13Rik|MGC22688|Serine/threonine kinase 32A|Serine/threonine protein kinase 32A|Serine/threonine-protein kinase 32A|ST32A_HUMAN|STK 32A|YANK 1|YANK1|Yet another novel kinase 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WU08
- Q8WU08
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together