Anti-PRAMEF17 Rabbit Recombinant Polyclonal Antibody
Catalog # 77795-202
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:PRAME family member 17 protein (69-259AA)
- Antigen symbol:PRAMEF17
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q5VTA0
- Antigen synonyms:PRAMEF17|PRAME family member 17
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: PRAMEF17
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human