Anti-CEBPE Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CEBPE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-226
- 77823-225
- Antigen symbol:
- CEBPE
- CEBPE
- Antigen name:
- CCAAT/enhancer-binding protein epsilon protein (1-200AA)
- CCAAT/enhancer-binding protein epsilon protein (1-200AA)
- Antigen synonyms:
- C/EBP epsilon| CCAAT/enhancer binding protein (C/EBP) epsilon| CCAAT/enhancer binding protein epsilon| CCAAT/enhancer-binding protein epsilon| CEBPE| CEBPE_HUMAN| CRP 1| CRP1
- C/EBP epsilon| CCAAT/enhancer binding protein (C/EBP) epsilon| CCAAT/enhancer binding protein epsilon| CCAAT/enhancer-binding protein epsilon| CEBPE| CEBPE_HUMAN| CRP 1| CRP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15744
- Q15744
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together