Anti-FABP3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FABP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77812-136
- 77812-135
- Antigen symbol:
- FABP3
- FABP3
- Antigen name:
- Fatty acid-binding protein, heart protein (2-133AA)
- Fatty acid-binding protein, heart protein (2-133AA)
- Antigen synonyms:
- 422 protein|Cardiac FABP|Cardiac Fatty Acid Binding Protein|FABP 11|FABP 3|FABP11|FABP3|FABPH_HUMAN|Fatty acid binding protein 11|Fatty acid binding protein 3|Fatty acid binding protein 3 muscle and heart|Fatty acid binding protein 3 muscle and heart mammary derived growth inhibitor|Fatty acid binding protein 3 muscle|muscle and heart (mammary derived growth inhibitor)|muscle|Fatty acid binding protein heart|Fatty acid binding protein|heart|muscle and heart|skeletal muscle|Fatty acid-binding protein 3|Fatty acid-binding protein|H FABP|H-FABP|Heart type fatty acid binding protein|Heart-type fatty acid-binding protein|M FABP|M-FABP|Mammary derived growth inhibitor|Mammary-derived growth inhibitor|MDGI|Muscle fatty acid binding protein|Muscle fatty acid-binding protein|Mylein protein P2 homolog|O FABP|OTTHUMP00000003898|P2 adopocyte protein
- 422 protein|Cardiac FABP|Cardiac Fatty Acid Binding Protein|FABP 11|FABP 3|FABP11|FABP3|FABPH_HUMAN|Fatty acid binding protein 11|Fatty acid binding protein 3|Fatty acid binding protein 3 muscle and heart|Fatty acid binding protein 3 muscle and heart mammary derived growth inhibitor|Fatty acid binding protein 3 muscle|muscle and heart (mammary derived growth inhibitor)|muscle|Fatty acid binding protein heart|Fatty acid binding protein|heart|muscle and heart|skeletal muscle|Fatty acid-binding protein 3|Fatty acid-binding protein|H FABP|H-FABP|Heart type fatty acid binding protein|Heart-type fatty acid-binding protein|M FABP|M-FABP|Mammary derived growth inhibitor|Mammary-derived growth inhibitor|MDGI|Muscle fatty acid binding protein|Muscle fatty acid-binding protein|Mylein protein P2 homolog|O FABP|OTTHUMP00000003898|P2 adopocyte protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P05413
- P05413
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together