Anti-CRTC3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CRTC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-104
- 77830-105
- Antigen symbol:
- CRTC3
- CRTC3
- Antigen name:
- CREB-regulated transcription coactivator 3 protein (373-480AA)
- CREB-regulated transcription coactivator 3 protein (373-480AA)
- Antigen synonyms:
- CREB regulated transcription coactivator 3|CREB-regulated transcription coactivator 3|CRTC 3||_HUMAN|FLJ21868|TORC 3|TORC-3|TORC3|Transducer of CREB protein 3|Transducer of regulated cAMP response element binding protein (CREB) 3|Transducer of regulated cAMP response element binding protein 3|Transducer of regulated cAMP response element-binding protein 3|Transducer of regulated CREB protein 3
- CREB regulated transcription coactivator 3|CREB-regulated transcription coactivator 3|CRTC 3||_HUMAN|FLJ21868|TORC 3|TORC-3|TORC3|Transducer of CREB protein 3|Transducer of regulated cAMP response element binding protein (CREB) 3|Transducer of regulated cAMP response element binding protein 3|Transducer of regulated cAMP response element-binding protein 3|Transducer of regulated CREB protein 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6UUV7
- Q6UUV7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together