Anti-DDX11 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: DDX11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-998
- 77832-999
- Antigen symbol:
- DDX11
- DDX11
- Antigen name:
- ATP-dependent DNA helicase DDX11 protein (1-220AA)
- ATP-dependent DNA helicase DDX11 protein (1-220AA)
- Antigen synonyms:
- CHL1| CHL1 related helicase gene 1| CHL1-like helicase homolog| CHL1-related protein 1| CHLR1| Ddx11| DDX11_HUMAN| DEAD/H (Asp Glu Ala Asp/His) box polypeptide 11| DEAD/H (Asp-Glu-Ala-Asp/His) box helicase 11| DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog| S. cerevisiae)| DEAD/H box| DEAD/H box protein 11| hCHLR1| Keratinocyte growth factor regulated gene 2 protein| Keratinocyte growth factor-regulated gene 2 protein| KRG 2| KRG-2| KRG2| MGC133249| MGC9335| OTTHUMP00000238778| OTTHUMP00000238780| OTTHUMP00000238784| OTTHUMP00000238785| Probable ATP dependent RNA helicase DDX11| Probable ATP-dependent RNA helicase DDX11| WABS
- CHL1| CHL1 related helicase gene 1| CHL1-like helicase homolog| CHL1-related protein 1| CHLR1| Ddx11| DDX11_HUMAN| DEAD/H (Asp Glu Ala Asp/His) box polypeptide 11| DEAD/H (Asp-Glu-Ala-Asp/His) box helicase 11| DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog| S. cerevisiae)| DEAD/H box| DEAD/H box protein 11| hCHLR1| Keratinocyte growth factor regulated gene 2 protein| Keratinocyte growth factor-regulated gene 2 protein| KRG 2| KRG-2| KRG2| MGC133249| MGC9335| OTTHUMP00000238778| OTTHUMP00000238780| OTTHUMP00000238784| OTTHUMP00000238785| Probable ATP dependent RNA helicase DDX11| Probable ATP-dependent RNA helicase DDX11| WABS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96FC9
- Q96FC9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together