Anti-pntB Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: pntB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli
Specifications
- Catalog No:
- 77817-760
- 77817-761
- Antigen symbol:
- pntB
- pntB
- Antigen name:
- Recombinant Escherichia coli NAD(P) transhydrogenase subunit beta protein (330-462AA)
- Recombinant Escherichia coli NAD(P) transhydrogenase subunit beta protein (330-462AA)
- Antigen synonyms:
- pntB|b1602|JW1594|NAD(P) transhydrogenase subunit beta|EC 7.1.1.1|Nicotinamide nucleotide transhydrogenase subunit beta|Pyridine nucleotide transhydrogenase subunit beta
- pntB|b1602|JW1594|NAD(P) transhydrogenase subunit beta|EC 7.1.1.1|Nicotinamide nucleotide transhydrogenase subunit beta|Pyridine nucleotide transhydrogenase subunit beta
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- E. coli
- E. coli
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P0AB67
- P0AB67
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together