Anti-PPP6R1 Rabbit Polyclonal Antibody
Catalog # 77798-654
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:SAPS1
- Antigen symbol:PPP6R1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9UPN7
- Antigen synonyms:PPP6R1|KIAA1115|PP6R1|SAPS1|Serine/threonine-protein phosphatase 6 regulatory subunit 1|SAPS domain family member 1
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PPP6R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat