Anti-SETD1A Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SETD1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77799-266
- 77799-267
- Antigen symbol:
- SETD1A
- SETD1A
- Antigen name:
- Histone-lysine N-methyltransferase SETD1A protein (188-304AA)
- Histone-lysine N-methyltransferase SETD1A protein (188-304AA)
- Antigen synonyms:
- Histone-lysine N-methyltransferase SETD1A|hSET1A|KMT2F|Lysine N methyltransferase 2F|Lysine N-methyltransferase 2F|SET domain containing 1A|SET domain containing protein 1A|SET domain-containing protein 1A|SET1|Set1 Ash2 histone methyltransferase complex subunit SET1|Set1/Ash2 histone methyltransferase complex subunit SET1|SET1A|SET1A_HUMAN
- Histone-lysine N-methyltransferase SETD1A|hSET1A|KMT2F|Lysine N methyltransferase 2F|Lysine N-methyltransferase 2F|SET domain containing 1A|SET domain containing protein 1A|SET domain-containing protein 1A|SET1|Set1 Ash2 histone methyltransferase complex subunit SET1|Set1/Ash2 histone methyltransferase complex subunit SET1|SET1A|SET1A_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15047
- O15047
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together