Anti-FKBP11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FKBP11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-991
- 77839-992
- Antigen symbol:
- FKBP11
- FKBP11
- Antigen name:
- Peptidyl-prolyl cis-trans isomerase FKBP11 protein (28-154AA)
- Peptidyl-prolyl cis-trans isomerase FKBP11 protein (28-154AA)
- Antigen synonyms:
- 19 kDa FK506 binding protein|19 kDa FK506-binding protein|19 kDa FKBP|FK506 binding protein 11 (19 kDa)|FK506 binding protein 11|FK506-binding protein 11|FKB11_HUMAN|FKBP 19 |FKBP-11|FKBP-19|Fkbp11|FKBP19 |MGC54182|Peptidyl prolyl cis trans isomerase|Peptidyl-prolyl cis-trans isomerase FKBP11|PPIase|PPIase FKBP11|Rotamase
- 19 kDa FK506 binding protein|19 kDa FK506-binding protein|19 kDa FKBP|FK506 binding protein 11 (19 kDa)|FK506 binding protein 11|FK506-binding protein 11|FKB11_HUMAN|FKBP 19 |FKBP-11|FKBP-19|Fkbp11|FKBP19 |MGC54182|Peptidyl prolyl cis trans isomerase|Peptidyl-prolyl cis-trans isomerase FKBP11|PPIase|PPIase FKBP11|Rotamase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NYL4
- Q9NYL4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together