Anti-DUSP19 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DUSP19
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-613
- 77833-614
- Antigen symbol:
- DUSP19
- DUSP19
- Antigen name:
- Dual specificity protein phosphatase 19 protein (1-217AA)
- Dual specificity protein phosphatase 19 protein (1-217AA)
- Antigen synonyms:
- Dual specificity phosphatase 19|Dual specificity phosphatase TS DSP1|Dual specificity phosphatase TS-DSP1|Dual specificity protein phosphatase 19|DUS19_HUMAN|DUSP 17|DUSP 19|DUSP17|LMW DSP3|LMW-DSP3|LMWDSP 3|LMWDSP3|Low molecular weight dual specificity phosphatase 3|MGC138210|Protein phosphatase SKRP1|SAPK pathway regulating phosphatase 1|SAPK pathway-regulating phosphatase 1|SKRP 1|SKRP1|Stress activated protein kinase pathway regulating phosphatase 1|Stress-activated protein kinase pathway-regulating phosphatase 1|TS DSP1
- Dual specificity phosphatase 19|Dual specificity phosphatase TS DSP1|Dual specificity phosphatase TS-DSP1|Dual specificity protein phosphatase 19|DUS19_HUMAN|DUSP 17|DUSP 19|DUSP17|LMW DSP3|LMW-DSP3|LMWDSP 3|LMWDSP3|Low molecular weight dual specificity phosphatase 3|MGC138210|Protein phosphatase SKRP1|SAPK pathway regulating phosphatase 1|SAPK pathway-regulating phosphatase 1|SKRP 1|SKRP1|Stress activated protein kinase pathway regulating phosphatase 1|Stress-activated protein kinase pathway-regulating phosphatase 1|TS DSP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WTR2
- Q8WTR2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together