Anti-RAB2A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RAB2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-387
- 77796-386
- Antigen symbol:
- RAB2A
- RAB2A
- Antigen name:
- Ras-related protein Rab-2A protein (2-212AA)
- Ras-related protein Rab-2A protein (2-212AA)
- Antigen synonyms:
- 9330148M11Rik|C80220|GTP binding protein|LHX|OTTMUSP00000004796|RAB2|RAB2 member RAS oncogene family|RAB2A|member RAS oncogene family|RAB2A_HUMAN|Ras related protein Rab 2A|RAS-associated protein RAB2|Ras-related protein Rab-2A|RP23-183L6.1|Small GTP binding protein RAB2A
- 9330148M11Rik|C80220|GTP binding protein|LHX|OTTMUSP00000004796|RAB2|RAB2 member RAS oncogene family|RAB2A|member RAS oncogene family|RAB2A_HUMAN|Ras related protein Rab 2A|RAS-associated protein RAB2|Ras-related protein Rab-2A|RP23-183L6.1|Small GTP binding protein RAB2A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61019
- P61019
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together