Anti-RAB5A Rabbit Recombinant Polyclonal Antibody
Catalog # 77796-450
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Ras-related protein Rab-5A protein (1-215AA)
- Antigen symbol:RAB5A
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P20339
- Antigen synonyms:RAB 5|RAB 5A|RAB5A|RAB5A member RAS oncogene family|RAB5A_HUMAN|RAS associated protein RAB5A|Ras related protein Rab 5A|Ras-related protein Rab-5A
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: RAB5A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat