Anti-KRTAP3-2 Rabbit Recombinant Polyclonal Antibody
77838-140
New Product
- Antigen name:Keratin-associated protein 3-2 protein (2-98AA)
- Antigen symbol:KRTAP3-2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BYR7
- Antigen synonyms:KAP3.2|KRTAP3.2Keratin-associated protein 3-2|High sulfur keratin-associated protein 3.2|Keratin-associated protein 3.2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: KRTAP3-2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human