Anti-KRT71 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KRT71
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-148
- 77826-149
- Antigen symbol:
- KRT71
- KRT71
- Antigen name:
- Keratin, type II cytoskeletal 71 protein (201-500AA)
- Keratin, type II cytoskeletal 71 protein (201-500AA)
- Antigen synonyms:
- CK-71| Cytokeratin-71| hK6irs| hK6irs1| K2C71_HUMAN| K6IRS1| K71| KB34| Keratin 6 irs| Keratin 6, inner root sheath, 1| Keratin| keratin, type II cytoskeletal 71| Keratin-71| KRT6IRS| KRT6IRS1| KRT71| MGC119390| MGC119391| OTTHUMP00000236990| type II cytoskeletal 71| Type II inner root sheath-specific keratin-K6irs1| Type-II keratin Kb34
- CK-71| Cytokeratin-71| hK6irs| hK6irs1| K2C71_HUMAN| K6IRS1| K71| KB34| Keratin 6 irs| Keratin 6, inner root sheath, 1| Keratin| keratin, type II cytoskeletal 71| Keratin-71| KRT6IRS| KRT6IRS1| KRT71| MGC119390| MGC119391| OTTHUMP00000236990| type II cytoskeletal 71| Type II inner root sheath-specific keratin-K6irs1| Type-II keratin Kb34
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q3SY84
- Q3SY84
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together