Anti-POU3F3 Rabbit Polyclonal Antibody
77794-898
New Product
- Antigen name:Synthetic peptide derived from POU3F3
- Antigen symbol:POU3F3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P20264
- Antigen synonyms:Brain 1|Brain specific homeobox/POU domain protein 1|Brain-1|Brain-specific homeobox/POU domain protein 1|Brain-specific homeobox/POU domain protein 1.1|Brain1|BRN 1|Brn-1|BRN1|Brn1, mouse, homolog of|class 3|Class III POU domain protein taichi|Oct-8|Octamer-binding protein 8|Octamer-binding transcription factor 8|OTF 8|OTF-8|OTF8|PO3 F3|PO3F 3|PO3F3|PO3F3_HUMAN|POU class 3 homeobox 3|POU domain|POU domain protein 12|POU domain, class 3, transcription factor 3|POU domain, class 3, transcription factor 3-A|pou12|POU3 F3|POU3F 3|Pou3f3|RHS 1|Rhs 2|RHS1|Rhs2|Skin 1|Skin1|tai-ji|transcription factor 3|zp12|zp12pou
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: POU3F3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat