Anti-CYCB1-1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: CYCB1-1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
- Catalog No:
- 77816-222
- 77816-221
- Antigen symbol:
- CYCB1-1
- CYCB1-1
- Antigen name:
- Recombinant Arabidopsis thaliana Cyclin-B1-1 protein (1-428AA)
- Recombinant Arabidopsis thaliana Cyclin-B1-1 protein (1-428AA)
- Antigen synonyms:
- CYCB1-1| CYC1| At4g37490| F6G17.140Cyclin-B1-1| Cyc1-At| G2/mitotic-specific cyclin-B1-1| CycB1|1
- CYCB1-1| CYC1| At4g37490| F6G17.140Cyclin-B1-1| Cyc1-At| G2/mitotic-specific cyclin-B1-1| CycB1|1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- A. thaliana
- A. thaliana
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P30183
- P30183
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together