Order Entry
Canada
ContactUsLinkComponent
 
Anti-ARL6IP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
 
undefined
Anti-ARL6IP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product

 

  • Catalog No:
  • 77822-597
  • 77822-596
  • Antigen symbol:
  • ARL6IP1
  • ARL6IP1
  • Antigen name:
  • ADP-ribosylation factor-like protein 6-interacting protein 1 protein (93-133AA)
  • ADP-ribosylation factor-like protein 6-interacting protein 1 protein (93-133AA)
  • Antigen synonyms:
  • ADP ribosylation factor like 6 interacting protein|ADP-ribosylation factor-like protein 6-interacting protein 1|Aip-1|Aip1|AR6P1_HUMAN|ARL-6-interacting protein 1|ARL6 interacting protein 1|ARL6IP|Arl6ip1|ARMER|KIAA0069
  • ADP ribosylation factor like 6 interacting protein|ADP-ribosylation factor-like protein 6-interacting protein 1|Aip-1|Aip1|AR6P1_HUMAN|ARL-6-interacting protein 1|ARL6 interacting protein 1|ARL6IP|Arl6ip1|ARMER|KIAA0069
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q15041
  • Q15041
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together