Anti-HOGA1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HOGA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-256
- 77832-257
- Antigen symbol:
- HOGA1
- HOGA1
- Antigen name:
- 4-hydroxy-2-oxoglutarate aldolase, mitochondrial protein (1-164AA)
- 4-hydroxy-2-oxoglutarate aldolase, mitochondrial protein (1-164AA)
- Antigen synonyms:
- HOGA1|C10orf65|DHDPSL|4-hydroxy-2-oxoglutarate aldolase|mitochondrial|EC 4.1.3.16|Dihydrodipicolinate synthase-like|DHDPS-like protein|Probable 2-keto-4-hydroxyglutarate aldolase|Probable KHG-aldolase|Protein 569272
- HOGA1|C10orf65|DHDPSL|4-hydroxy-2-oxoglutarate aldolase|mitochondrial|EC 4.1.3.16|Dihydrodipicolinate synthase-like|DHDPS-like protein|Probable 2-keto-4-hydroxyglutarate aldolase|Probable KHG-aldolase|Protein 569272
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86XE5
- Q86XE5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together