Anti-SYNGAP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SYNGAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-299
- 77834-300
- Antigen symbol:
- SYNGAP1
- SYNGAP1
- Antigen name:
- Ras/Rap GTPase-activating protein SynGAP protein (1161-1343AA)
- Ras/Rap GTPase-activating protein SynGAP protein (1161-1343AA)
- Antigen synonyms:
- DKFZp761G1421|KIAA1938|MRD5|Neuronal RasGAP|OTTHUMP00000064825|p135 SynGAP|Ras GTPase activating protein SynGAP|Ras GTPase-activating protein SynGAP|RASA 1|RASA 5|RASA1|RASA5|SYGP1_HUMAN|Synaptic Ras GAP 1|Synaptic Ras GTPase activating protein 1|Synaptic Ras GTPase activating protein 1 homolog|Synaptic Ras GTPase activating protein 135kDa|Synaptic Ras GTPase activating protein|Synaptic Ras GTPase-activating protein 1|Synaptic Ras-GAP 1|SYNGAP 1|SYNGAP1
- DKFZp761G1421|KIAA1938|MRD5|Neuronal RasGAP|OTTHUMP00000064825|p135 SynGAP|Ras GTPase activating protein SynGAP|Ras GTPase-activating protein SynGAP|RASA 1|RASA 5|RASA1|RASA5|SYGP1_HUMAN|Synaptic Ras GAP 1|Synaptic Ras GTPase activating protein 1|Synaptic Ras GTPase activating protein 1 homolog|Synaptic Ras GTPase activating protein 135kDa|Synaptic Ras GTPase activating protein|Synaptic Ras GTPase-activating protein 1|Synaptic Ras-GAP 1|SYNGAP 1|SYNGAP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96PV0
- Q96PV0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together