Anti-GCKR Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GCKR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-844
- 77824-843
- Antigen symbol:
- GCKR
- GCKR
- Antigen name:
- Glucokinase regulatory protein (1-625AA)
- Glucokinase regulatory protein (1-625AA)
- Antigen synonyms:
- FGQTL5|GCK|GCKR_HUMAN|GKRP|GLRE|glucokinase (hexokinase 4) regulator|Glucokinase|Glucokinase regulator|Glucokinase regulatory protein|Hexokinase 4 regulator|Hexokinase type IV|Hexokinase-4|Hexokinase-D|HK IV|HK4|OTTHUMP00000158533
- FGQTL5|GCK|GCKR_HUMAN|GKRP|GLRE|glucokinase (hexokinase 4) regulator|Glucokinase|Glucokinase regulator|Glucokinase regulatory protein|Hexokinase 4 regulator|Hexokinase type IV|Hexokinase-4|Hexokinase-D|HK IV|HK4|OTTHUMP00000158533
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14397
- Q14397
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together