Order Entry
Canada
ContactUsLinkComponent
Anti-SARS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-SARS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-SARS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77807-532
  • 77807-533
  • Antigen symbol:
  • SARS
  • SARS
  • Antigen name:
  • Serine--tRNA ligase, cytoplasmic protein (2-233AA)
  • Serine--tRNA ligase, cytoplasmic protein (2-233AA)
  • Antigen synonyms:
  • C78314|cytoplasmic|EC 6.1.1.11|FLJ36399|sarS|Sars1|Serine tRNA ligase 1, cytoplasmic|Serine tRNA ligase|Serine--tRNA ligase|serine--tRNA ligase, cytoplasmic|SerRS|SERS|Seryl tRNA Ser/Sec synthetase|Seryl tRNA synthetase|Seryl tRNA synthetase cytoplasmic|Seryl-tRNA Ser/Sec synthetase|Seryl-tRNA synthetase|seryl-tRNA synthetase, cytoplasmic|Seryl-tRNA(Ser/Sec) synthetase|Strs|SYSC_HUMAN
  • C78314|cytoplasmic|EC 6.1.1.11|FLJ36399|sarS|Sars1|Serine tRNA ligase 1, cytoplasmic|Serine tRNA ligase|Serine--tRNA ligase|serine--tRNA ligase, cytoplasmic|SerRS|SERS|Seryl tRNA Ser/Sec synthetase|Seryl tRNA synthetase|Seryl tRNA synthetase cytoplasmic|Seryl-tRNA Ser/Sec synthetase|Seryl-tRNA synthetase|seryl-tRNA synthetase, cytoplasmic|Seryl-tRNA(Ser/Sec) synthetase|Strs|SYSC_HUMAN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P49591
  • P49591
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together