Anti-TRMT112 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRMT112
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-459
- 77841-460
- Antigen symbol:
- TRMT112
- TRMT112
- Antigen name:
- tRNA methyltransferase 112 homolog protein (1-125AA)
- tRNA methyltransferase 112 homolog protein (1-125AA)
- Antigen synonyms:
- TRMT112| AD-001| HSPC152| HSPC170| Multifunctional methyltransferase subunit TRM112-like protein| tRNA methyltransferase 112 homolog
- TRMT112| AD-001| HSPC152| HSPC170| Multifunctional methyltransferase subunit TRM112-like protein| tRNA methyltransferase 112 homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UI30
- Q9UI30
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together