Anti-VGLL1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: VGLL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77834-634
- 77834-635
- Antigen symbol:
- VGLL1
- VGLL1
- Antigen name:
- Transcription cofactor vestigial-like protein 1 protein (139-250AA)
- Transcription cofactor vestigial-like protein 1 protein (139-250AA)
- Antigen synonyms:
- Protein TONDU| TDU| TONDU| Transcription cofactor vestigial like protein 1| Transcription cofactor vestigial-like protein 1| Vestigial like 1| vestigial like 1 homolog (Drosophila)| Vgl-1| VGL1| Vgll1| VGLL1_HUMAN
- Protein TONDU| TDU| TONDU| Transcription cofactor vestigial like protein 1| Transcription cofactor vestigial-like protein 1| Vestigial like 1| vestigial like 1 homolog (Drosophila)| Vgl-1| VGL1| Vgll1| VGLL1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99990
- Q99990
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together