Anti-PXK Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PXK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-115
- 77832-116
- Antigen symbol:
- PXK
- PXK
- Antigen name:
- PX domain-containing protein kinase-like protein (1-200AA)
- PX domain-containing protein kinase-like protein (1-200AA)
- Antigen synonyms:
- FLJ20335|K-ATPase|Modulator of Na|Modulator of Na K ATPase|Modulator of Na K ATPase long form|MONaKA|PX domain containing protein kinase like protein|PX domain containing serine/threonine kinase|PX domain-containing protein kinase-like protein|PX ser/thr kinase v2|PX serine/threonine kinase|Pxk|PXK_HUMAN
- FLJ20335|K-ATPase|Modulator of Na|Modulator of Na K ATPase|Modulator of Na K ATPase long form|MONaKA|PX domain containing protein kinase like protein|PX domain containing serine/threonine kinase|PX domain-containing protein kinase-like protein|PX ser/thr kinase v2|PX serine/threonine kinase|Pxk|PXK_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z7A4
- Q7Z7A4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together