Order Entry
Canada
ContactUsLinkComponent
Anti-RHOD Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Anti-RHOD Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-RHOD Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77797-319
  • 77797-318
  • Antigen symbol:
  • RHOD
  • RHOD
  • Antigen name:
  • Rho-related GTP-binding protein RhoD protein (1-80AA)
  • Rho-related GTP-binding protein RhoD protein (1-80AA)
  • Antigen synonyms:
  • ARHD|Ras homolog D|Ras homolog gene family member A|Ras homolog gene family member D|Rho|RHO D|Rho related GTP binding protein RhoD|Rho related protein HP1|Rho-related GTP-binding protein RhoD|Rho-related protein HP1|RHOD|RHOD_HUMAN|RhoHP1|RHOM
  • ARHD|Ras homolog D|Ras homolog gene family member A|Ras homolog gene family member D|Rho|RHO D|Rho related GTP binding protein RhoD|Rho related protein HP1|Rho-related GTP-binding protein RhoD|Rho-related protein HP1|RHOD|RHOD_HUMAN|RhoHP1|RHOM
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • O00212
  • O00212
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together