Anti-SEMA4F Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: SEMA4F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77831-292
- 77831-293
- Antigen symbol:
- SEMA4F
- SEMA4F
- Antigen name:
- Fusion protein of SEMA4F
- Fusion protein of SEMA4F
- Antigen synonyms:
- M SEMA| m Sema M| PRO2353| SEM4F_HUMAN| Sema domain immunoglobulin domain (Ig) transmembrane| Sema M| Sema W| Sema4f| SEMAM| Semaphorin M| Semaphorin W| Semaphorin-4F| Semaphorin-M| Semaphorin-W| SEMAW
- M SEMA| m Sema M| PRO2353| SEM4F_HUMAN| Sema domain immunoglobulin domain (Ig) transmembrane| Sema M| Sema W| Sema4f| SEMAM| Semaphorin M| Semaphorin W| Semaphorin-4F| Semaphorin-M| Semaphorin-W| SEMAW
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95754
- O95754
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together