Anti-CARD10 Rabbit Recombinant Polyclonal Antibody
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CARD10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77838-063
- 77838-062
- Antigen symbol:
- CARD10
- CARD10
- Antigen name:
- Caspase recruitment domain-containing protein 10 protein (863-1032AA)
- Caspase recruitment domain-containing protein 10 protein (863-1032AA)
- Antigen synonyms:
- Bcl10 binding protein and activator of NFKB|BIMP1|CAR10_HUMAN|CARD 10|CARD containing MAGUK 3 protein|CARD-containing MAGUK 3|CARD-containing MAGUK protein 3|CARD10|Carma 3|CARMA3|Caspase Recruitment Domain family member 10|Caspase recruitment domain-containing protein 10
- Bcl10 binding protein and activator of NFKB|BIMP1|CAR10_HUMAN|CARD 10|CARD containing MAGUK 3 protein|CARD-containing MAGUK 3|CARD-containing MAGUK protein 3|CARD10|Carma 3|CARMA3|Caspase Recruitment Domain family member 10|Caspase recruitment domain-containing protein 10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BWT7
- Q9BWT7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together