Anti-TRAF3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRAF3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-448
- 77803-448
- Antigen symbol:
- TRAF3
- TRAF3
- Antigen name:
- TNF receptor-associated factor 3 protein (1-40AA)
- TNF receptor-associated factor 3 protein (1-40AA)
- Antigen synonyms:
- CAP 1|CAP-1|CAP1|CD40 associated protein 1|CD40 binding protein|CD40 bp|CD40 receptor associated factor 1|CD40 receptor-associated factor 1|CD40-binding protein|CD40BP|CRAF 1|CRAF1|IIAE5|LAP 1|LAP1|LMP1 associated protein|LMP1-associated protein 1|MIPT3|TNF receptor associated factor 3|TNF receptor-associated factor 3|TRAF 3|Traf3|TRAF3_HUMAN
- CAP 1|CAP-1|CAP1|CD40 associated protein 1|CD40 binding protein|CD40 bp|CD40 receptor associated factor 1|CD40 receptor-associated factor 1|CD40-binding protein|CD40BP|CRAF 1|CRAF1|IIAE5|LAP 1|LAP1|LMP1 associated protein|LMP1-associated protein 1|MIPT3|TNF receptor associated factor 3|TNF receptor-associated factor 3|TRAF 3|Traf3|TRAF3_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13114
- Q13114
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together