Anti-CCL4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CCL4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77809-050
- 77809-049
- Antigen symbol:
- CCL4
- CCL4
- Antigen name:
- C-C motif chemokine 4 protein (24-84AA)
- C-C motif chemokine 4 protein (24-84AA)
- Antigen synonyms:
- MIP 1 beta| Secreted protein G 26|ACT 2|ACT-2|ACT2|AT744.1|AT744.2|C C motif chemokine 4|C C motif chemokine 4 like|C C motif chemokine ligand 4 like 1|C C motif chemokine ligand 4 like 2|CC chemokine ligand 4|CC chemokine ligand 4L1|CC chemokine ligand 4L1d2|CC chemokine ligand 4L2|CCL4_HUMAN|ccl4l 1|CCL4L|CCL4L1|Chemokine (C C motif) ligand 4|Chemokine (C C motif) ligand 4 like 1|telomeric|Chemokine (C C motif) ligand 4 like 2|Chemokine CC Motif Ligand 4|G 26|G 26 T lymphocyte secreted protein|G-26 T-lymphocyte-secreted protein|HC21|Immune activation 2|LAG 1|LAG-1|LAG1|Lymphocyte activation gene 1|Lymphocyte activation gene 1 protein|Macrophage inflammatory protein 1 beta|Macrophage inflammatory protein 1-beta|Macrophage inflammatory protein 1b2|MGC104418|MGC126025|MGC126026|MIP-1-beta|MIP-1-beta(1-69)|MIP-1-beta(3-69)|MIP1 beta|MIP1B|MIP1B1|Monocyte adherence induced protein 5 alpha|PAT 744|Protein H400|SCYA2|SCYA4|SCYA4L|SCYA4L1|SCYA4L2|SCYQ4L2|Secreted protein G 26|Secreted protein G26|SIS gamma|SIS-gamma|Small inducible cytokine A4 (homologous to mouse Mip 1b)|Small inducible cytokine A4|small inducible cytokine A4-like|Small-inducible cytokine A4|T cell activation protein 2|T-cell activation protein 2
- MIP 1 beta| Secreted protein G 26|ACT 2|ACT-2|ACT2|AT744.1|AT744.2|C C motif chemokine 4|C C motif chemokine 4 like|C C motif chemokine ligand 4 like 1|C C motif chemokine ligand 4 like 2|CC chemokine ligand 4|CC chemokine ligand 4L1|CC chemokine ligand 4L1d2|CC chemokine ligand 4L2|CCL4_HUMAN|ccl4l 1|CCL4L|CCL4L1|Chemokine (C C motif) ligand 4|Chemokine (C C motif) ligand 4 like 1|telomeric|Chemokine (C C motif) ligand 4 like 2|Chemokine CC Motif Ligand 4|G 26|G 26 T lymphocyte secreted protein|G-26 T-lymphocyte-secreted protein|HC21|Immune activation 2|LAG 1|LAG-1|LAG1|Lymphocyte activation gene 1|Lymphocyte activation gene 1 protein|Macrophage inflammatory protein 1 beta|Macrophage inflammatory protein 1-beta|Macrophage inflammatory protein 1b2|MGC104418|MGC126025|MGC126026|MIP-1-beta|MIP-1-beta(1-69)|MIP-1-beta(3-69)|MIP1 beta|MIP1B|MIP1B1|Monocyte adherence induced protein 5 alpha|PAT 744|Protein H400|SCYA2|SCYA4|SCYA4L|SCYA4L1|SCYA4L2|SCYQ4L2|Secreted protein G 26|Secreted protein G26|SIS gamma|SIS-gamma|Small inducible cytokine A4 (homologous to mouse Mip 1b)|Small inducible cytokine A4|small inducible cytokine A4-like|Small-inducible cytokine A4|T cell activation protein 2|T-cell activation protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P13236
- P13236
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together