Anti-SHARPIN Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SHARPIN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77837-066
- 77837-065
- Antigen symbol:
- SHARPIN
- SHARPIN
- Antigen name:
- Sharpin protein (82-171AA)
- Sharpin protein (82-171AA)
- Antigen synonyms:
- DKFZp434N1923|hSIPL1|Shank associated RH domain interacting protein|SHANK associated RH domain interactor|Shank interacting protein like 1|Shank-associated RH domain-interacting protein|Shank-interacting protein-like 1|Sharpin|SHRPN_HUMAN|SIPL1
- DKFZp434N1923|hSIPL1|Shank associated RH domain interacting protein|SHANK associated RH domain interactor|Shank interacting protein like 1|Shank-associated RH domain-interacting protein|Shank-interacting protein-like 1|Sharpin|SHRPN_HUMAN|SIPL1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H0F6
- Q9H0F6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together