Anti-CPPED1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CPPED1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-945
- 77837-944
- Antigen symbol:
- CPPED1
- CPPED1
- Antigen name:
- Serine/threonine-protein phosphatase CPPED1 protein (49-186AA)
- Serine/threonine-protein phosphatase CPPED1 protein (49-186AA)
- Antigen synonyms:
- CPPED1|CSTP1Serine/threonine-protein phosphatase CPPED1|EC 3.1.3.16|Calcineurin-like phosphoesterase domain-containing protein 1|Complete S-transactivated protein 1
- CPPED1|CSTP1Serine/threonine-protein phosphatase CPPED1|EC 3.1.3.16|Calcineurin-like phosphoesterase domain-containing protein 1|Complete S-transactivated protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BRF8
- Q9BRF8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together