Anti-STIP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: STIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-349
- 77801-348
- Antigen symbol:
- STIP1
- STIP1
- Antigen name:
- Stress-induced-phosphoprotein 1 protein (1-543AA)
- Stress-induced-phosphoprotein 1 protein (1-543AA)
- Antigen synonyms:
- Epididymis secretory sperm binding protein Li 94n|HEL S 94n|Hop|Hsc70/Hsp90 organizing protein|Hsc70/Hsp90-organizing protein|IEF SSP 3521|NY REN 11 antigen|P60|Renal carcinoma antigen NY-REN-11|STI1|STI1L|STIP1_HUMAN|Stress induced phosphoprotein 1|Stress-induced-phosphoprotein 1|Transformation sensitive protein IEF SSP 3521|Transformation-sensitive protein IEF SSP 3521
- Epididymis secretory sperm binding protein Li 94n|HEL S 94n|Hop|Hsc70/Hsp90 organizing protein|Hsc70/Hsp90-organizing protein|IEF SSP 3521|NY REN 11 antigen|P60|Renal carcinoma antigen NY-REN-11|STI1|STI1L|STIP1_HUMAN|Stress induced phosphoprotein 1|Stress-induced-phosphoprotein 1|Transformation sensitive protein IEF SSP 3521|Transformation-sensitive protein IEF SSP 3521
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31948
- P31948
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together