Anti-PARP2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77836-769
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Poly [ADP-ribose] polymerase 2 protein (14-149AA)
- Antigen symbol:PARP2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UGN5
- Antigen synonyms:ADP ribosyltransferase like 2|ADP-ribosyltransferase diphtheria toxin-like 2|ADPRT 2|ADPRT-2|ADPRT2|ADPRTL 2|ADPRTL 3|ADPRTL2|ADPRTL3|ARTD2|hPARP 2|hPARP-2|hPARP2|NAD(+) ADP ribosyltransferase 2|NAD(+) ADP-ribosyltransferase 2|pADPRT 2|pADPRT-2|pADPRT2|PARP 2|PARP-2|PARP2|PARP2_HUMAN|Poly (ADP ribose) polymerase family member 2|Poly (ADP ribosyl) transferase like 2|poly (ADP-ribose) polymerase 2|Poly [ADP ribose] synthetase 2|Poly [ADP-ribose] polymerase 2|Poly(ADP ribose) synthetase|Poly[ADP-ribose] synthase 2
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: PARP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human