Anti-PIGQ Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIGQ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-639
- 77841-640
- Antigen symbol:
- PIGQ
- PIGQ
- Antigen name:
- Phosphatidylinositol N-acetylglucosaminyltransferase subunit Q protein (616-760AA)
- Phosphatidylinositol N-acetylglucosaminyltransferase subunit Q protein (616-760AA)
- Antigen synonyms:
- Phosphatidylinositol glycan, class Q|c407A10.1|GPI1|hGPI1|MGC12693|N-acetylglucosamyl transferase component GPI1|Phosphatidylinositol glycan anchor biosynthesis, class Q|Phosphatidylinositol N acetylglucosaminyltransferase subunit Q|Phosphatidylinositol N-acetylglucosaminyltransferase subunit Q|Phosphatidylinositol-glycan biosynthesis class Q protein|PIG Q|PIG-Q|PIGQ|PIGQ_HUMAN
- Phosphatidylinositol glycan, class Q|c407A10.1|GPI1|hGPI1|MGC12693|N-acetylglucosamyl transferase component GPI1|Phosphatidylinositol glycan anchor biosynthesis, class Q|Phosphatidylinositol N acetylglucosaminyltransferase subunit Q|Phosphatidylinositol N-acetylglucosaminyltransferase subunit Q|Phosphatidylinositol-glycan biosynthesis class Q protein|PIG Q|PIG-Q|PIGQ|PIGQ_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BRB3
- Q9BRB3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together