Anti-ATP8A2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ATP8A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-496
- 77836-497
- Antigen symbol:
- ATP8A2
- ATP8A2
- Antigen name:
- Phospholipid-transporting ATPase IB protein (358-570AA)
- Phospholipid-transporting ATPase IB protein (358-570AA)
- Antigen synonyms:
- AT8A2_HUMAN| ATP| Atp8a2| ATPase aminophospholipid transporter class I type 8A member 2| ATPase aminophospholipid transporter like class I type 8A member 2| ATPase class I type 8A member 2| ATPase phospholipid transporting 8A2| ATPIB| CAMRQ4| EC=3.6.3.1| IB| ML-1| ML1| P4 ATPase flippase complex alpha subunit ATP8A2| Probable phospholipid-transporting ATPase IB
- AT8A2_HUMAN| ATP| Atp8a2| ATPase aminophospholipid transporter class I type 8A member 2| ATPase aminophospholipid transporter like class I type 8A member 2| ATPase class I type 8A member 2| ATPase phospholipid transporting 8A2| ATPIB| CAMRQ4| EC=3.6.3.1| IB| ML-1| ML1| P4 ATPase flippase complex alpha subunit ATP8A2| Probable phospholipid-transporting ATPase IB
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NTI2
- Q9NTI2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together