Anti-FBXW10 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FBXW10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-639
- 77828-640
- Antigen symbol:
- FBXW10
- FBXW10
- Antigen name:
- F-box/WD repeat-containing protein 10 protein (701-1000AA)
- F-box/WD repeat-containing protein 10 protein (701-1000AA)
- Antigen synonyms:
- FBXW10|F-box/WD repeat-containing protein 10|F-box and WD-40 domain-containing protein 10|Ubiquitin ligase-specificity factor
- FBXW10|F-box/WD repeat-containing protein 10|F-box and WD-40 domain-containing protein 10|Ubiquitin ligase-specificity factor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5XX13
- Q5XX13
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together