Order Entry
Canada
ContactUsLinkComponent
Anti-KIF24 Rabbit Recombinant Polyclonal Antibody
Anti-KIF24 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-KIF24 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77827-991
  • 77827-992
  • Antigen symbol:
  • KIF24
  • KIF24
  • Antigen name:
  • Kinesin-like protein KIF24 protein (1168-1368AA)
  • Kinesin-like protein KIF24 protein (1168-1368AA)
  • Antigen synonyms:
  • bA571F15.4|C9orf48|KIF24_HUMAN|kinesin family member 24|Kinesin-like protein RP11-571F15.7
  • bA571F15.4|C9orf48|KIF24_HUMAN|kinesin family member 24|Kinesin-like protein RP11-571F15.7
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q5T7B8
  • Q5T7B8
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together