Anti-PECR Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PECR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77793-691
- 77793-690
- Antigen symbol:
- PECR
- PECR
- Antigen name:
- Peroxisomal trans-2-enoyl-CoA reductase protein (24-303AA)
- Peroxisomal trans-2-enoyl-CoA reductase protein (24-303AA)
- Antigen synonyms:
- 2 4 dienoyl CoA reductase related protein|2|4-dienoyl-CoA reductase-related protein|DCR RP|DCR-RP|DCRRP|HPDHase|HSA250303|OTTHUMP00000206786|PECR|PECR_HUMAN|Peroxisomal trans 2 enoyl CoA reductase|Peroxisomal trans-2-enoyl-CoA reductase|PRO1004|Putative short chain alcohol dehydrogenase|pVI ARL|pVI-ARL|PVIARL|SDR29C1|Short chain dehydrogenase/reductase family 29C member 1|TERP
- 2 4 dienoyl CoA reductase related protein|2|4-dienoyl-CoA reductase-related protein|DCR RP|DCR-RP|DCRRP|HPDHase|HSA250303|OTTHUMP00000206786|PECR|PECR_HUMAN|Peroxisomal trans 2 enoyl CoA reductase|Peroxisomal trans-2-enoyl-CoA reductase|PRO1004|Putative short chain alcohol dehydrogenase|pVI ARL|pVI-ARL|PVIARL|SDR29C1|Short chain dehydrogenase/reductase family 29C member 1|TERP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BY49
- Q9BY49
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together