Anti-SASH1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SASH1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77798-666
- 77798-665
- Antigen symbol:
- SASH1
- SASH1
- Antigen name:
- SAM and SH3 domain-containing protein 1 protein (76-191AA)
- SAM and SH3 domain-containing protein 1 protein (76-191AA)
- Antigen synonyms:
- 2500002E12Rik| dJ323M4| dJ323M4.1| KIAA0790| PEPE 1| PEPE1| Proline glutamate repeat containing protein| Proline-glutamate repeat-containing protein| RP3 323M4.1| SAM and SH3 domain containing 1| SAM and SH3 domain containing protein 1| SAM and SH3 domain-containing protein 1| SASH 1| Sash1| SASH1_HUMAN
- 2500002E12Rik| dJ323M4| dJ323M4.1| KIAA0790| PEPE 1| PEPE1| Proline glutamate repeat containing protein| Proline-glutamate repeat-containing protein| RP3 323M4.1| SAM and SH3 domain containing 1| SAM and SH3 domain containing protein 1| SAM and SH3 domain-containing protein 1| SASH 1| Sash1| SASH1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O94885
- O94885
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together