Anti-OBSCN Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: OBSCN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-781
- 77828-782
- Antigen symbol:
- OBSCN
- OBSCN
- Antigen name:
- Obscurin protein (1621-1712AA)
- Obscurin protein (1621-1712AA)
- Antigen synonyms:
- BC046431|Gm878|KIAA1556|KIAA1639||_HUMAN|Obscurin|Obscurin-MLCK|Obscurin-myosin light chain kinase|Obscurin-RhoGEF|OTTMUSG00000005786|UNC89
- BC046431|Gm878|KIAA1556|KIAA1639||_HUMAN|Obscurin|Obscurin-MLCK|Obscurin-myosin light chain kinase|Obscurin-RhoGEF|OTTMUSG00000005786|UNC89
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5VST9
- Q5VST9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together