Anti-MAP3K7CL Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: MAP3K7CL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77816-680
- 77816-679
- Antigen symbol:
- MAP3K7CL
- MAP3K7CL
- Antigen name:
- MAP3K7 C-terminal-like protein (1-142AA)
- MAP3K7 C-terminal-like protein (1-142AA)
- Antigen synonyms:
- MAP3K7CL|C21orf7|TAK1LMAP3K7 C-terminal-like protein|TAK1-like protein
- MAP3K7CL|C21orf7|TAK1LMAP3K7 C-terminal-like protein|TAK1-like protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P57077
- P57077
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together