Anti-PQBP1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PQBP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-181
- 77795-180
- Antigen symbol:
- PQBP1
- PQBP1
- Antigen name:
- Polyglutamine-binding protein 1 protein (2-265AA)
- Polyglutamine-binding protein 1 protein (2-265AA)
- Antigen synonyms:
- 38 kDa nuclear protein containing a WW domain|Mental retardation|X linked 55|MRX55|MRXS3|MRXS8|Npw38|Nuclear protein containing WW domain 38 kD|OTTHUMP00000025808|Polyglutamine binding protein 1|Polyglutamine tract binding protein 1|Polyglutamine tract-binding protein 1|Polyglutamine-binding protein 1|PQBP 1|PQBP-1|PQBP1|PQBP1_HUMAN|RENS1|SHS|Sutherland Haan X linked mental retardation syndrome
- 38 kDa nuclear protein containing a WW domain|Mental retardation|X linked 55|MRX55|MRXS3|MRXS8|Npw38|Nuclear protein containing WW domain 38 kD|OTTHUMP00000025808|Polyglutamine binding protein 1|Polyglutamine tract binding protein 1|Polyglutamine tract-binding protein 1|Polyglutamine-binding protein 1|PQBP 1|PQBP-1|PQBP1|PQBP1_HUMAN|RENS1|SHS|Sutherland Haan X linked mental retardation syndrome
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60828
- O60828
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together