Anti-HMGXB4 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HMGXB4
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-924
- 77837-923
- Antigen symbol:
- HMGXB4
- HMGXB4
- Antigen name:
- HMG domain-containing protein 4 protein (489-509AA)
- HMG domain-containing protein 4 protein (489-509AA)
- Antigen synonyms:
- HMGXB4|HMG2L1|HMGBCG|HMG domain-containing protein 4|HMG box-containing protein 4|High mobility group protein 2-like 1|Protein HMGBCG
- HMGXB4|HMG2L1|HMGBCG|HMG domain-containing protein 4|HMG box-containing protein 4|High mobility group protein 2-like 1|Protein HMGBCG
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UGU5
- Q9UGU5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together