Anti-OCIAD1 Rabbit Recombinant Antibody (Biotinylated)
New Product
Type:
Antigen: OCIAD1
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-074
- 77841-075
- Antigen symbol:
- OCIAD1
- OCIAD1
- Antigen name:
- OCIA domain-containing protein 1 protein (1-245AA)
- OCIA domain-containing protein 1 protein (1-245AA)
- Antigen synonyms:
- Asrij|FLJ20455|MGC111072|OCAD1_HUMAN|OCIA|OCIA domain containing 1|OCIA domain-containing protein 1|OCIAD1|Ovarian carcinoma immunoreactive antigen|TPA018
- Asrij|FLJ20455|MGC111072|OCAD1_HUMAN|OCIA|OCIA domain containing 1|OCIA domain-containing protein 1|OCIAD1|Ovarian carcinoma immunoreactive antigen|TPA018
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NX40
- Q9NX40
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together